ATC359510198 Atlantic Research Chemicals Ltd

Methyl Quinoxalin-6-ylacetate

  • MDL number MFCD19706065
  • Cas Number : 1233318-23-4
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES02803
Availability details for
Quantity
  • Availability
  • Add to Cart