ATC359510210 Atlantic Research Chemicals Ltd

Methyl 1-(2-Methoxy-1-methylethyl)-5-oxopyrrolidine-3-carboxylate

  • MDL number MFCD19706060
  • Cas Number : 1355334-47-2
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES04518
Availability details for
Quantity
  • Availability
  • Add to Cart