ATC359510226 Atlantic Research Chemicals Ltd

Methyl (4-Oxocyclohexyl)acetate

  • MDL number MFCD09031995
  • Cas Number : 66405-41-2
  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES09936
Availability details for
Quantity
  • Availability
  • Add to Cart