ATC359510300 Atlantic Research Chemicals Ltd

2-Chloro-3-(trifluoromethyl)benzyl alcohol

  • MDL number MFCD01631537
  • Cas Number : 261763-20-6
  • Molecular Weight : 210.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=HM00137
Availability details for
Quantity
  • Availability
  • Add to Cart