ATC359510312 Atlantic Research Chemicals Ltd

3-(6-Methylpyridin-2-yl)propan-1-ol

  • MDL number MFCD00012344
  • Cas Number : 61744-43-2
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HM00230
Availability details for
Quantity
  • Availability
  • Add to Cart