ATC359510331 Atlantic Research Chemicals Ltd

4-(Chloromethyl)-2-phenyl-1,3-thiazole

  • MDL number MFCD00723503
  • Cas Number : 4771-31-7
  • Molecular Weight : 209.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=MH00607
Availability details for
Quantity
  • Availability
  • Add to Cart