ATC359510395 Atlantic Research Chemicals Ltd

2-({[(4-Methylphenyl)sulphonyl]oxy}amino)ethanamine

  • MDL number MFCD00276588
  • Cas Number : 175205-36-4
  • Molecular Weight : 230.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001704
Availability details for
Quantity
  • Availability
  • Add to Cart