ATC359510410 Atlantic Research Chemicals Ltd

N-(2,4-Dinitrophenyl)-N-methylamine

  • MDL number MFCD00024399
  • Cas Number : 2044-88-4
  • Molecular Weight : 197.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE001221
Availability details for
Quantity
  • Availability
  • Add to Cart