ATC359510434 Atlantic Research Chemicals Ltd

2,6-Difluorobenzohydrazide

  • MDL number MFCD00119406
  • Cas Number : 172935-91-0
  • Molecular Weight : 172.133

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=XS001743
Availability details for
Quantity
  • Availability
  • Add to Cart