ATC359510443 Atlantic Research Chemicals Ltd

5-Fluoro-2,6-dihydroxynicotinonitrile

  • MDL number MFCD03001419
  • Cas Number : 113237-18-6
  • Molecular Weight : 154.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002328
Availability details for
Quantity
  • Availability
  • Add to Cart