ATC359510449 Atlantic Research Chemicals Ltd

N-Cyclopentyl-3-nitropyridin-2-amine

  • MDL number MFCD08692157
  • Cas Number : 952934-74-6
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002334
Availability details for
Quantity
  • Availability
  • Add to Cart