ATC359510456 Atlantic Research Chemicals Ltd

tert-Butyl 4-(3-Nitropyridin-2-yl)piperazine-1-carboxylate

  • MDL number MFCD09028514
  • Cas Number : 153473-24-6
  • Molecular Weight : 308.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002341
Availability details for
Quantity
  • Availability
  • Add to Cart