ATC359510469 Atlantic Research Chemicals Ltd

2-Amino-5-iodonicotinonitrile

  • MDL number MFCD20040120
  • Cas Number : 1347815-41-1
  • Molecular Weight : 245.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002354
Availability details for
Quantity
  • Availability
  • Add to Cart