ATC359510490 Atlantic Research Chemicals Ltd

Tetrahydrofuran-2-carboxamide

  • MDL number MFCD03929553
  • Cas Number : 91470-28-9
  • Molecular Weight : 115.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS00426
Availability details for
Quantity
  • Availability
  • Add to Cart