ATC359510525 Atlantic Research Chemicals Ltd

5-Methyl-2H-1,4-benzoxazin-3(4H)-one

  • MDL number MFCD12755935
  • Cas Number : 31366-49-1
  • Molecular Weight : 163.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS02913
Availability details for
Quantity
  • Availability
  • Add to Cart