ATC359510536 Atlantic Research Chemicals Ltd

Piperidinepentanenitrile

  • MDL number MFCD00044836
  • Cas Number : 4734-49-0
  • Molecular Weight : 166.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS03796
Availability details for
Quantity
  • Availability
  • Add to Cart