ATC359510543 Atlantic Research Chemicals Ltd

1-(2-Chlorobenzyl)piperazine

  • MDL number MFCD01860843
  • Cas Number : 17532-19-3
  • Molecular Weight : 210.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS03869
Availability details for
Quantity
  • Availability
  • Add to Cart