ATC359510582 Atlantic Research Chemicals Ltd

1,3-Dihydro-2H-imidazo[4,5-b]pyridin-2-one

  • MDL number MFCD00464845
  • Cas Number : 16328-62-4
  • Molecular Weight : 135.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS100207
Availability details for
Quantity
  • Availability
  • Add to Cart