ATC386058193 Atlantic Research Chemicals Ltd

Neopentylamine

  • MDL number MFCD00008134
  • Cas Number : 5813-64-9
  • Molecular Weight : 87.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE10124
Availability details for
Quantity
  • Availability
  • Add to Cart