ATC386058202 Atlantic Research Chemicals Ltd

3-(4-Nitrophenoxy)benzaldehyde

  • MDL number MFCD18207479
  • Cas Number : 17076-72-1
  • Molecular Weight : 243.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AH001188
Availability details for
Quantity
  • Availability
  • Add to Cart