ATC386058261 Atlantic Research Chemicals Ltd

(1S)-1-(4-Pyridyl)ethylamine

  • MDL number MFCD09256819
  • Cas Number : 27854-96-2
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AM00257
Availability details for
Quantity
  • Availability
  • Add to Cart