ATC386058316 Atlantic Research Chemicals Ltd

4-(4-Ethylphenyl)-4-oxobutanoic acid

  • MDL number MFCD00173766
  • Cas Number : 49594-75-4
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001537
Availability details for
Quantity
  • Availability
  • Add to Cart