ATC386058318 Atlantic Research Chemicals Ltd

4-Oxo-4-(4-propoxyphenyl)butanoic acid

  • MDL number MFCD00100661
  • Cas Number : 39496-82-7
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001539
Availability details for
Quantity
  • Availability
  • Add to Cart