ATC386058331 Atlantic Research Chemicals Ltd

3-Chlorophenoxyacetic acid�

  • MDL number MFCD00015850
  • Molecular Weight : 186.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA001552
Availability details for
Quantity
  • Availability
  • Add to Cart