ATC386058372 Atlantic Research Chemicals Ltd

4-[(2,2-Dimethylpropanoyl)amino]nicotinic acid

  • MDL number MFCD03407344
  • Cas Number : 125867-31-4
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA002393
Availability details for
Quantity
  • Availability
  • Add to Cart