ATC386058398 Atlantic Research Chemicals Ltd

5-(Benzyloxy)-1H-indole-2-carboxylic acid

  • MDL number MFCD00047165
  • Cas Number : 6640-09-1
  • Molecular Weight : 267.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA01921
Availability details for
Quantity
  • Availability
  • Add to Cart