ATC386058470 Atlantic Research Chemicals Ltd

3-(4-Chlorophenyl)acryloyl chloride

  • MDL number MFCD00039313
  • Cas Number : 35086-79-4
  • Molecular Weight : 201.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CC00146
Availability details for
Quantity
  • Availability
  • Add to Cart