ATC386058479 Atlantic Research Chemicals Ltd

4-Bromo-2,6-difluorobenzoyl chloride

  • MDL number MFCD03094086
  • Cas Number : 497181-19-8
  • Molecular Weight : 255.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CC00155
Availability details for
Quantity
  • Availability
  • Add to Cart