ATC386058484 Atlantic Research Chemicals Ltd

2-Methyl-3-(trifluoromethyl)benzoyl chloride

  • MDL number MFCD01631598
  • Cas Number : 261952-07-2
  • Molecular Weight : 222.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CC00160
Availability details for
Quantity
  • Availability
  • Add to Cart