ATC386058541 Atlantic Research Chemicals Ltd

1H-Benzimidazole-2-carbonyl chloride

  • MDL number MFCD16987526
  • Cas Number : 30183-14-3
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CC02201
Availability details for
Quantity
  • Availability
  • Add to Cart