ATC386058555 Atlantic Research Chemicals Ltd

1-Methyl-1-cyclohexanecarbonyl chloride

  • MDL number MFCD00019346
  • Cas Number : 2890-61-1
  • Molecular Weight : 160.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CC09923
Availability details for
Quantity
  • Availability
  • Add to Cart