ATC386058560 Atlantic Research Chemicals Ltd

4,4-Difluorocyclohexanecarbonyl chloride

  • MDL number MFCD13173983
  • Cas Number : 376348-75-3
  • Molecular Weight : 182.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CC09928
Availability details for
Quantity
  • Availability
  • Add to Cart