ATC386058570 Atlantic Research Chemicals Ltd

Methyl 2-(4-bromophenyl)-2,2-dimethylacetate

  • MDL number MFCD02683357
  • Cas Number : 154825-97-5
  • Molecular Weight : 257.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES001174
Availability details for
Quantity
  • Availability
  • Add to Cart