ATC386058580 Atlantic Research Chemicals Ltd

Ethyl 2,6-Difluorobenzoate

  • MDL number MFCD06204338
  • Cas Number : 19064-14-3
  • Molecular Weight : 186.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES001184
Availability details for
Quantity
  • Availability
  • Add to Cart