ATC386058599 Atlantic Research Chemicals Ltd

Methyl (4-Nitrophenyl)acetate

  • MDL number MFCD00024802
  • Cas Number : 2945-08-6
  • Molecular Weight : 195.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES001203
Availability details for
Quantity
  • Availability
  • Add to Cart