ATC386058621 Atlantic Research Chemicals Ltd

Methyl 2-Chloroisonicotinate 1-oxide

  • MDL number MFCD14582313
  • Cas Number : 164464-50-0
  • Molecular Weight : 187.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES002109
Availability details for
Quantity
  • Availability
  • Add to Cart