ATC386058632 Atlantic Research Chemicals Ltd

Methyl Pyridin-3-ylacetate

  • MDL number MFCD00023617
  • Cas Number : 39998-25-9
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES002120
Availability details for
Quantity
  • Availability
  • Add to Cart