ATC386058646 Atlantic Research Chemicals Ltd

Methyl 2-Oxooxazolidine-4-carboxylate

  • MDL number MFCD21609325
  • Cas Number : 96751-61-0
  • Molecular Weight : 145.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00811
Availability details for
Quantity
  • Availability
  • Add to Cart