ATC386058670 Atlantic Research Chemicals Ltd

Methyl 5-Oxopyrrolidine-3-carboxylate

  • MDL number MFCD08275678
  • Cas Number : 35309-35-4
  • Molecular Weight : 143.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES04521
Availability details for
Quantity
  • Availability
  • Add to Cart