ATC386058685 Atlantic Research Chemicals Ltd

Methyl Cyclopentylacetate

  • MDL number MFCD12031703
  • Cas Number : 2723-38-8
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES09954
Availability details for
Quantity
  • Availability
  • Add to Cart