ATC386058688 Atlantic Research Chemicals Ltd

Ethyl (6-Amino-9H-purin-9-yl)acetate

  • MDL number MFCD01321238
  • Cas Number : 25477-96-7
  • Molecular Weight : 221.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES10065
Availability details for
Quantity
  • Availability
  • Add to Cart