ATC386058739 Atlantic Research Chemicals Ltd

4-(3-Chloro-2-pyridinyl)morpholine

  • MDL number MFCD00498941
  • Cas Number : 54231-36-6
  • Molecular Weight : 198.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA002260
Availability details for
Quantity
  • Availability
  • Add to Cart