ATC386058741 Atlantic Research Chemicals Ltd

2,6-Dichloro-3-methylpyridine

  • MDL number MFCD00137847
  • Cas Number : 58584-94-4
  • Molecular Weight : 162.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA002262
Availability details for
Quantity
  • Availability
  • Add to Cart