ATC386058857 Atlantic Research Chemicals Ltd

N-Benzyl-N-(4-nitrophenyl)amine

  • MDL number MFCD00126504
  • Cas Number : 14309-92-3
  • Molecular Weight : 228.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001757
Availability details for
Quantity
  • Availability
  • Add to Cart