ATC386058883 Atlantic Research Chemicals Ltd

2-Chlorobenzene-1-carbohydrazide

  • MDL number MFCD0000759
  • Cas Number : 5814-05-1
  • Molecular Weight : 170.598

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=XS001783
Availability details for
Quantity
  • Availability
  • Add to Cart