ATC386058890 Atlantic Research Chemicals Ltd

4-(Benzylamino)phenol

  • MDL number MFCD00051312
  • Cas Number : 103-14-0
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001790
Availability details for
Quantity
  • Availability
  • Add to Cart