ATC386058896 Atlantic Research Chemicals Ltd

4-(Pyridin-2-yloxy)benzonitrile

  • MDL number MFCD08543445
  • Cas Number : 270260-33-8
  • Molecular Weight : 196.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001797
Availability details for
Quantity
  • Availability
  • Add to Cart