ATC386058931 Atlantic Research Chemicals Ltd

1-(4-Chlorophenyl)-2-(4-methylphenyl)ethane-1,2-dione

  • MDL number MFCD00052718
  • Cas Number : 86508-29-4
  • Molecular Weight : 258.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001836
Availability details for
Quantity
  • Availability
  • Add to Cart