ATC386058932 Atlantic Research Chemicals Ltd

N'-Hydroxy-4-(4-methoxyphenoxy)-3-nitrobenzenecarboximidamide

  • MDL number MFCD00662654
  • Cas Number : 261966-14-7
  • Molecular Weight : 303.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001837
Availability details for
Quantity
  • Availability
  • Add to Cart