ATC386058942 Atlantic Research Chemicals Ltd

N1-[4-(Trifluoromethoxy)phenyl]acetamide

  • MDL number MFCD00075293
  • Cas Number : 1737-06-0
  • Molecular Weight : 219.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS001848
Availability details for
Quantity
  • Availability
  • Add to Cart